ENA410542771 Enamine

2-[(1-methyl-1H-indol-3-yl)formamido]hexanoic acid

  • MDL number MFCD13706058

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-137693

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-137693
Availability details for
Quantity
  • Availability
  • Add to Cart