ENA410542772 Enamine

3-{cyclopropyl[(2-methoxyphenyl)carbamoyl]amino}propanoic acid

  • MDL number MFCD26407883
  • Cas Number : 1178064-88-4
  • Molecular Weight : 278.31

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-137694

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-137694
Availability details for
Quantity
  • Availability
  • Add to Cart