ENA410542773 Enamine

3-{[(2-methoxypyridin-3-yl)formamido]methyl}benzoic acid

  • MDL number MFCD26407884
  • Cas Number : 1179647-14-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-137695

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-137695
Availability details for
Quantity
  • Availability
  • Add to Cart