ENA410542781 Enamine

1-(2-methoxyethyl)-2,3-dihydro-1H-isoindole

  • MDL number MFCD21136821
  • Cas Number : 127797-17-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Ice Pack
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-137739

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-137739
Availability details for
Quantity
  • Availability
  • Add to Cart