ENA410542784 Enamine

(2R)-2-amino-3-methylbutanenitrile hydrochloride

  • MDL number MFCD26407891
  • Cas Number : 1798890-45-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-137770

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-137770
Availability details for
Quantity
  • Availability
  • Add to Cart