ENA410542789 Enamine

2-(2-chloro-6-methoxyphenyl)acetic acid

  • MDL number MFCD18393596
  • Cas Number : 170737-94-7
  • Molecular Weight : 200.62

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name (2-chloro-6-methoxyphenyl)acetic acid
Inchi Code 1S/C9H9ClO3/c1-13-8-4-2-3-7(10)6(8)5-9(11)12/h2-4H,5H2,1H3,(H,11,12)
Inchi Key DVHQMZULSWDQIF-UHFFFAOYSA-N

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-137777

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-137777
Availability details for
Quantity
  • Availability
  • Add to Cart