ENA410542800 Enamine

6-bromo-2,3,4,5-tetrahydro-1H-2-benzazepine hydrochloride

  • MDL number MFCD26407893
  • Cas Number : 1803609-55-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 254...256
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-137793

Safety Information

msds https://www.enaminestore.com/catalog/EN300-137793
Availability details for
Quantity
  • Availability
  • Add to Cart