ENA410542808 Enamine

3-{[(3-methylphenyl)methyl](phenyl)amino}propanoic acid

  • MDL number MFCD26407900
  • Cas Number : 1179771-60-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form oil
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-137843

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-137843
Availability details for
Quantity
  • Availability
  • Add to Cart