ENA410542809 Enamine

methyl 5-(2-fluorophenoxymethyl)-1,2-oxazole-3-carboxylate

  • MDL number MFCD10485481
  • Cas Number : 932780-86-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 73...75
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-137848

Safety Information

msds https://www.enaminestore.com/catalog/EN300-137848
Availability details for
Quantity
  • Availability
  • Add to Cart