ENA410542819 Enamine

3-(4-chlorophenyl)-4-methyl-4H-1,2,4-triazole

  • MDL number MFCD03175175
  • Cas Number : 168968-40-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-137909

Safety Information

msds https://www.enaminestore.com/catalog/EN300-137909
Availability details for
Quantity
  • Availability
  • Add to Cart