ENA410542820 Enamine

2-[(2,5-dimethylphenyl)sulfanyl]acetonitrile

  • MDL number MFCD04622354
  • Cas Number : 103575-52-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-07030

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-07030
Availability details for
Quantity
  • Availability
  • Add to Cart