ENA410542829 Enamine

2-(2,6-dimethylpiperidin-1-yl)-2-oxoacetic acid

  • MDL number MFCD12171729
  • Cas Number : 768329-98-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-138009

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-138009
Availability details for
Quantity
  • Availability
  • Add to Cart