ENA410542830 Enamine

3-amino-1-(4-bromo-3-methylphenyl)piperidin-2-one

  • MDL number MFCD19679412
  • Cas Number : 1343710-09-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-138010

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-138010
Availability details for
Quantity
  • Availability
  • Add to Cart