ENA410542831 Enamine

3-cyclobutanecarbonyl-5-nitro-1H-indole

  • MDL number MFCD11545710
  • Cas Number : 1097117-03-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-138011

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-138011
Availability details for
Quantity
  • Availability
  • Add to Cart