ENA410542832 Enamine

1,3-dimethyl 2-(4-bromophenyl)-2-methylpropanedioate

  • MDL number MFCD26407913
  • Cas Number : 243835-94-1
  • Molecular Weight : 301.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form liquid
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-138012

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-138012
Availability details for
Quantity
  • Availability
  • Add to Cart