ENA410542844 Enamine

1-[2-(benzyloxy)-4-bromophenyl]ethan-1-one

  • MDL number MFCD25579578
  • Cas Number : 161385-97-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-138068

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-138068
Availability details for
Quantity
  • Availability
  • Add to Cart