ENA410542855 Enamine

3-(hydroxymethyl)pyridine-2-carboxamide

  • MDL number MFCD18802493
  • Cas Number : 115012-12-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-138160

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-138160
Availability details for
Quantity
  • Availability
  • Add to Cart