ENA410542857 Enamine

2-(cyclopentylsulfanyl)-6-fluorobenzoic acid

  • MDL number MFCD21944952
  • Cas Number : 1540844-88-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 78...80
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-138164

Safety Information

msds https://www.enaminestore.com/catalog/EN300-138164
Availability details for
Quantity
  • Availability
  • Add to Cart