ENA410542858 Enamine

4-chloro-2-(cyclopentylsulfanyl)benzoic acid

  • MDL number MFCD18270817
  • Cas Number : 1343726-37-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-138165

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-138165
Availability details for
Quantity
  • Availability
  • Add to Cart