ENA410542861 Enamine

1-(4-fluorophenyl)-4-methyl-1H-pyrazol-3-amine

  • MDL number MFCD16845442
  • Cas Number : 1251278-62-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 106...108
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-138169

Safety Information

msds https://www.enaminestore.com/catalog/EN300-138169
Availability details for
Quantity
  • Availability
  • Add to Cart