ENA410542862 Enamine

2-(3-methylpyrrolidin-1-yl)-2-oxoacetic acid

  • MDL number MFCD14601463
  • Cas Number : 1251118-37-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-138170

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-138170
Availability details for
Quantity
  • Availability
  • Add to Cart