ENA410542863 Enamine

(4-chloro-3-fluorophenyl)(1H-1,2,3,4-tetrazol-5-yl)methanamine

  • MDL number MFCD16040268
  • Cas Number : 1247472-41-8
  • Molecular Weight : 227.63

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-138178

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-138178
Availability details for
Quantity
  • Availability
  • Add to Cart