ENA410542867 Enamine

1-(2-bromo-4-methoxyphenyl)-2-(4-methylphenyl)ethan-1-one

  • MDL number MFCD12517686
  • Cas Number : 1183068-56-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 81...83
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-138184

Safety Information

msds https://www.enaminestore.com/catalog/EN300-138184
Availability details for
Quantity
  • Availability
  • Add to Cart