ENA410542875 Enamine

ethyl 6-(4-chlorophenoxy)hexanoate

  • MDL number MFCD20156644
  • Cas Number : 1410382-45-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-138226

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-138226
Availability details for
Quantity
  • Availability
  • Add to Cart