ENA410542888 Enamine

4-bromo-1-(methoxymethyl)-3,5-dimethyl-1H-pyrazole

  • MDL number MFCD18324055
  • Cas Number : 1263210-45-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-101705

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-101705
Availability details for
Quantity
  • Availability
  • Add to Cart