ENA410542893 Enamine

3-(benzyloxy)cyclobutan-1-amine hydrochloride

  • MDL number MFCD16657503
  • Cas Number : 92146-76-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 166...168
Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-138282

Safety Information

msds https://www.enaminestore.com/catalog/EN300-138282
Availability details for
Quantity
  • Availability
  • Add to Cart