ENA410542897 Enamine

2-(2-fluoro-3-hydroxyphenyl)acetic acid

  • MDL number MFCD09263180
  • Cas Number : 1213229-25-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-138287

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-138287
Availability details for
Quantity
  • Availability
  • Add to Cart