ENA410542900 Enamine

ethyl 5-ethoxy-1,3-oxazole-2-carboxylate

  • MDL number MFCD07787333
  • Cas Number : 68208-09-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-138300

Safety Information

signalword Warning
msds https://www.enaminestore.com/catalog/EN300-138300
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart