ENA410542908 Enamine

2,2-difluoro-3-hydroxy-3-methylbutanoic acid

  • MDL number MFCD16749721
  • Cas Number : 15080-23-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-138310

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-138310
Availability details for
Quantity
  • Availability
  • Add to Cart