ENA410542913 Enamine

4-phenyl-2,3-dihydro-1H-indole hydrochloride

  • MDL number MFCD26407935
  • Cas Number : 1797090-67-5
  • Molecular Weight : 231.72

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

mp 216-217
Iupac Name 4-phenylindoline hydrochloride
Inchi Code 1S/C14H13N.ClH/c1-2-5-11(6-3-1)12-7-4-8-14-13(12)9-10-15-14;/h1-8,15H,9-10H2;1H
Inchi Key DMKYMOBRXNNGGN-UHFFFAOYSA-N

Description

Melting Point 216-217
Purity 95
Salt Data Cl
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-138344

Safety Information

signalword Warning
pcodes P261,P264,P270,P271,P280,P301+P312,P302+P352,P304+P340,P305+P351+P338,P312,P330,P332+P313,P337+P313,P362,P403+P233,P405,P501
MSDS https://www.enaminestore.com/catalog/EN300-138344
pictograms GHS07
hstatements H302,H315,H319,H335
Availability details for
Quantity
  • Availability
  • Add to Cart