ENA410542917 Enamine

N-(2-aminocyclohexyl)-4-butoxybenzamide hydrochloride

  • MDL number MFCD26407936
  • Cas Number : 1607262-19-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-138351

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-138351
Availability details for
Quantity
  • Availability
  • Add to Cart