ENA410542921 Enamine

5-bromo-3,3,7-trimethyl-2,3-dihydro-1H-indol-2-one

  • MDL number MFCD16657839
  • Cas Number : 1379363-44-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 180...182
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-138549

Safety Information

msds https://www.enaminestore.com/catalog/EN300-138549
Availability details for
Quantity
  • Availability
  • Add to Cart