ENA410542923 Enamine

1-(2-bromo-4-methoxyphenyl)-2-(2-methylphenyl)ethan-1-one

  • MDL number MFCD12517679
  • Cas Number : 1183611-53-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-138551

Safety Information

msds https://www.enaminestore.com/catalog/EN300-138551
Availability details for
Quantity
  • Availability
  • Add to Cart