ENA410542924 Enamine

4-(chloromethyl)-5-(trifluoromethyl)-1H-pyrazole

  • MDL number MFCD21774601
  • Molecular Weight : 184.55

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 4-(chloromethyl)-5-(trifluoromethyl)-1H-pyrazole
Inchi Code 1S/C5H4ClF3N2/c6-1-3-2-10-11-4(3)5(7,8)9/h2H,1H2,(H,10,11)
Inchi Key AGBBKAQHIOKOFJ-UHFFFAOYSA-N

Description

Purity 95
Storage temp. +4
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-138552

Safety Information

signalword Danger
pcodes P261,P264,P270,P271,P280,P301+P312,P304+P340,P305+P351+P338,P310,P312,P330,P501
MSDS https://www.enaminestore.com/catalog/EN300-138552
pictograms GHS05,GHS07
hstatements H302,H318,H332
Availability details for
Quantity
  • Availability
  • Add to Cart