ENA410542928 Enamine

1-[5-(1-benzothiophen-5-yl)furan-2-yl]ethan-1-one

  • MDL number MFCD21527888
  • Cas Number : 1490665-95-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-138559

Safety Information

msds https://www.enaminestore.com/catalog/EN300-138559
Availability details for
Quantity
  • Availability
  • Add to Cart