ENA410542929 Enamine

1-(5-bromopyridin-2-yl)-3-(propan-2-yl)urea

  • MDL number MFCD01833022
  • Cas Number : 353259-08-2
  • Molecular Weight : 258.12

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-138560

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-138560
Availability details for
Quantity
  • Availability
  • Add to Cart