ENA410542931 Enamine

methyl 6-(pyrrolidin-1-yl)pyridazine-3-carboxylate

  • MDL number MFCD09057801
  • Cas Number : 927963-87-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 187...189
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-138572

Safety Information

msds https://www.enaminestore.com/catalog/EN300-138572
Availability details for
Quantity
  • Availability
  • Add to Cart