ENA410542933 Enamine

methyl 3-(naphthalen-2-yl)propanoate

  • MDL number MFCD22381365
  • Cas Number : 81711-55-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 57...59
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-138575

Safety Information

msds https://www.enaminestore.com/catalog/EN300-138575
Availability details for
Quantity
  • Availability
  • Add to Cart