ENA410542934 Enamine

N-(4-amino-2-fluorophenyl)-3-chlorobenzamide

  • MDL number MFCD16499710
  • Cas Number : 1283396-12-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 119...121
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-138580

Safety Information

msds https://www.enaminestore.com/catalog/EN300-138580
Availability details for
Quantity
  • Availability
  • Add to Cart