ENA410542944 Enamine

methyl 1-bromocyclopentane-1-carboxylate

  • MDL number MFCD13192148
  • Cas Number : 51572-54-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-138596

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-138596
Availability details for
Quantity
  • Availability
  • Add to Cart