ENA410542945 Enamine

1-bromocyclopentane-1-carbonyl chloride

  • MDL number MFCD21602079
  • Cas Number : 56447-23-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. -20
Shipping Temperature Ice Pack
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-138597

Safety Information

msds https://www.enaminestore.com/catalog/EN300-138597
Availability details for
Quantity
  • Availability
  • Add to Cart