ENA410542949 Enamine

4-chloro-2-methylpyrazolo[1,5-a]pyrazine

  • MDL number MFCD11045717
  • Cas Number : 1314928-61-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-138646

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-138646
Availability details for
Quantity
  • Availability
  • Add to Cart