ENA410542951 Enamine

1-(oxiran-2-yl)ethan-1-ol

  • MDL number MFCD19300625
  • Cas Number : 765-44-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. -20
Shipping Temperature Ice Pack
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-138664

Safety Information

signalword Danger
pcodes P261,P264,P270,P271,P280,P301+P310,P302+P352,P304+P340,P311,P312,P330,P361,P363,P403+P233,P405,P501
MSDS https://www.enaminestore.com/catalog/EN300-138664
pictograms GHS06
hstatements H301,H311,H331
Availability details for
Quantity
  • Availability
  • Add to Cart