ENA410542956 Enamine

2-[(2,2-dimethoxyethyl)amino]ethan-1-ol

  • MDL number MFCD18865474
  • Cas Number : 98961-86-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form liquid
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-103016

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-103016
Availability details for
Quantity
  • Availability
  • Add to Cart