ENA410542962 Enamine

1-(3,5-dichlorophenyl)-2-methylpropan-1-ol

  • MDL number MFCD19602045
  • Cas Number : 1341140-47-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-138680

Safety Information

msds https://www.enaminestore.com/catalog/EN300-138680
Availability details for
Quantity
  • Availability
  • Add to Cart