ENA410542963 Enamine

1-(bromomethyl)-4-fluoro-5-methoxy-2-nitrobenzene

  • MDL number MFCD20084854
  • Cas Number : 1342850-43-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 71...73
Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-138681

Safety Information

msds https://www.enaminestore.com/catalog/EN300-138681
Availability details for
Quantity
  • Availability
  • Add to Cart