ENA410542964 Enamine

[2-(1,3-benzothiazol-2-yl)ethenyl]dimethylamine

  • MDL number MFCD26407945
  • Cas Number : 20973-89-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 92...94
Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-138686

Safety Information

msds https://www.enaminestore.com/catalog/EN300-138686
Availability details for
Quantity
  • Availability
  • Add to Cart