ENA410542965 Enamine

N-{[5-(pyrrolidin-2-yl)furan-2-yl]methyl}acetamide

  • MDL number MFCD26407946
  • Cas Number : 1713602-56-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-138688

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-138688
Availability details for
Quantity
  • Availability
  • Add to Cart