ENA410542966 Enamine

1-(3-bromopropanesulfonyl)-3-methylbenzene

  • MDL number MFCD11615282
  • Cas Number : 1082893-96-6
  • Molecular Weight : 277.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 1-[(3-bromopropyl)sulfonyl]-3-methylbenzene
Inchi Code 1S/C10H13BrO2S/c1-9-4-2-5-10(8-9)14(12,13)7-3-6-11/h2,4-5,8H,3,6-7H2,1H3
Inchi Key WMLPDQRPGWRUQE-UHFFFAOYSA-N

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-138689

Safety Information

signalword Warning
pcodes P261,P264,P270,P271,P280,P301+P312,P302+P352,P304+P340,P305+P351+P338,P312,P330,P332+P313,P337+P313,P362,P363,P403+P233,P405,P501
MSDS https://www.enaminestore.com/catalog/EN300-138689
pictograms GHS07
hstatements H302,H312,H315,H319,H332,H335
Availability details for
Quantity
  • Availability
  • Add to Cart