ENA410542969 Enamine

5-bromo-1-cyclopentyl-1H-pyrazolo[3,4-b]pyridine

  • MDL number MFCD09933349
  • Cas Number : 1016771-98-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-138692

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-138692
Availability details for
Quantity
  • Availability
  • Add to Cart